Learn More
Medchemexpress LLC Benzyl-PEG45-alcohol | 26403-74-7 | C97H188O46 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15315350MG
Encompass_Preferred
Benzyl-PEG45-alcohol is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Consists of two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets the protein of interest
Catalog No. 50-003-73446