Learn More
Medchemexpress LLC Benzyl-PEG5-acid | 2514948-41-3 | 356.41 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1384871G
Encompass_Preferred
Benzyl-PEG5-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. These compounds contain two different ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a protein of interest. PROTACs utilize the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-51016