Learn More
Medchemexpress LLC Benzyl-PEG5-acid | 2514948-41-3 | 356.41 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138487100MG
Encompass_Preferred
Benzyl-PEG5-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is intended for research use only and is not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-003-48629