Learn More
Medchemexpress LLC Benzyl-PEG5-acid | 2514948-41-3 | 356.41 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY138487
Encompass_Preferred
Benzyl-PEG5-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two distinct ligands with a linker, one targeting an E3 ubiquitin ligase and the other the protein of interest.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51851