Learn More
Medchemexpress LLC Benzyl-PEG5-Ots 500mg
SDP
Supplier:  Medchemexpress LLC HYW108220500MG
Encompass_Preferred
Benzyl-PEG5-Ots is a PEG-based PROTAC linker can be used in the synthesis of PROTACs
Catalog No. 50-002-05507