Learn More
Medchemexpress LLC Benzyl-PEG5-THP 25mg
SDP
Supplier:  Medchemexpress LLC 25MGHYW096075
Encompass_Preferred
Benzyl-PEG5-THP is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-06562