Learn More
Medchemexpress LLC Benzyl-PEG6-alcohol | 24342-68-5 | MFCD06797107 | 99.9% | 372.45 | C19H32O7 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0426545G
Encompass_Preferred
BnO-PEG6-OH is a benzyl-protected six-unit polyethylene glycol linker used as a non-cleavable spacer in antibody-drug conjugate (ADC) construction and as a PEG-based PROTAC linker. It provides a flexible, hydrophilic spacer for conjugation chemistry and is supplied at high purity for research applications.
  • Non-cleavable six-unit PEG linker suitable for spacer applications.
  • Applicable to ADC and PROTAC synthesis for flexible linkage.
  • Molecular formula C19H32O7 and molecular weight 372.45.
  • High purity suitable for research use.
  • Available in multiple pack sizes, including 5 G.
Catalog No. 50-002-03230