Learn More
Medchemexpress LLC Benzyl-PEG9-alcohol 1g
SDP
Supplier:  Medchemexpress LLC HYW0938331G
Encompass_Preferred
Benzyl-PEG9-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05928