Learn More
Medchemexpress LLC Benzyl-PEG9-alcohol 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW093833
Encompass_Preferred
Benzyl-PEG9-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04571