Learn More
Medchemexpress LLC Benzyl-PEG9-alcohol 500mg
SDP
Supplier:  Medchemexpress LLC HYW093833500MG
Encompass_Preferred
Benzyl-PEG9-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05890