Learn More
Medchemexpress LLC Benzyl tetrahydro-1H-pyrrolo[3,4-c]isoxazole-5(3H)-carboxylate | 1174730-77-8 | 95.0% | C13H16N2O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY75071500MG
Encompass_Preferred
Benzyl tetrahydro-1H-pyrrolo[3,4-c]isoxazole-5(3H)-carboxylate is a drug intermediate employed in the synthesis of various active compounds. It is supplied as a liquid with a light brown to brown color.
  • Used as a building block for complex molecules.
  • Available in multiple quantities for research needs.
  • Stored at -20°C for up to 3 years in pure form.
  • Detailed data sheet and safety data available.
Catalog No. 50-004-34386