Learn More
Selleck Chemical LLC Benzylamiloride hydrochloride S0406-5MG
SDP
Supplier:  Selleck Chemical LLC S04065MG
Encompass_Preferred
Benzylamiloride hydrochloride (Benzamil hydrochloride) is an inhibitor of Na /Ca2 exchanger (NCX) with an IC50 100 nM It reduces hyponatremic brain edema by blocking sodium influx in vivo
Catalog No. 50-000-43338