Learn More
Medchemexpress LLC Benzylboronic acid pinacol ester | 87100-28-5 | MFCD05663841 | 218.10 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00099310MM/1ML
Encompass_Preferred
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It finds application in Pd (0)-mediated [11 C] carbonylation reactions.
  • Biochemical reagent
  • Used as a biological material or organic compound
  • Useful for life science related research
  • Applied in Pd (0)-mediated [11 C] carbonylation reactions
Catalog No. 50-004-49023