Learn More
Medchemexpress LLC Benzyldimethyltetradecylammonium-d7 (chloride) | 1219178-72-9 | 95.1% | 375.08 | C23H35D7ClN | 5 MG

Supplier: Medchemexpress LLC HY132643S5MG
Benzyldimethyltetradecylammonium-d7 (chloride) is the deuterium-labeled isotopologue of benzyldimethyltetradecylammonium chloride supplied as the chloride salt. It is intended for research use as a stable isotope-labeled reference standard or tracer in mass spectrometry and analytical workflows. Solid, off-white powder with reported purity of 95.1%.
- Deuterium-labeled quaternary ammonium chloride for use as an internal standard.
- Suitable for mass spectrometry and tracer studies.
- Molecular formula C23H35D7ClN and molecular weight 375.08.
- Supplied as a solid powder with reported purity of 95.1%.
- Recommended storage: 4°C sealed; in solvent, -80°C up to 6 months or -20°C up to 1 month.
- CAS number 1219178-72-9 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.