Learn More
Medchemexpress LLC Benzyloxy-C5-PEG1 50mg
SDP
Supplier:  Medchemexpress LLC HYW09613450MG
Encompass_Preferred
Benzyloxy-C5-PEG1 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05721