Learn More
Medchemexpress LLC Benzyloxy carbonyl-PEG3-C2-acid | 2100306-73-6 | C17H24O7 | 1 G
SDP
Supplier:  Medchemexpress LLC HY14074550MG
Encompass_Preferred
Benzyloxy carbonyl-PEG3-C2-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is applied in cancer-programmed cell death research. This product has a molecular weight of 340.37 and is soluble at 10 mM in DMSO.
  • Used in the synthesis of PROTACs.
  • Applied in cancer-programmed cell death research.
  • Molecular weight of 340.37.
  • Soluble at 10 mM in DMSO.
  • Targets PROTAC linkers.
Catalog No. 50-003-54965