Learn More
Medchemexpress LLC Benzyloxy carbonyl-PEG3-C2-acid | 2100306-73-6 | C17H24O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140745100MG
Encompass_Preferred
Benzyloxy carbonyl-PEG3-C2-acid is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and is applicable in cancer-programmed cell death research. The compound has a molecular weight of 340.37 and is soluble at 10 mM in DMSO.
  • PEG-based PROTAC linker
  • Utilized in synthesis of PROTACs
  • Applicable in cancer-programmed cell death research
  • Molecular weight of 340.37
  • Soluble at 10 mM in DMSO
  • Targets PROTAC linkers
Catalog No. 50-003-56478