Learn More
Medchemexpress LLC Benzyloxycarbonyl-alanyl-glutamyl-valyl-aspartic acid fluoromethyl ketone | 1135688-47-9 | MFCD03453601 | 95.0% | 610.63 g/mol | C28H39FN4O10 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12530510MG
Encompass_Preferred
Z-AEVD-FMK is a peptide fluoromethyl ketone inhibitor of caspase-10 used in apoptosis research. It is cell-permeable and inhibits caspase-mediated cleavage events, reducing Bid activation and release of apoptosis-inducing factor (AIF) from mitochondria. The compound is provided as a solid with high purity and is intended for biochemical and cell-based assays; handle with appropriate safety precautions and prepare stock solutions in DMSO.
Catalog No. 50-002-37774