Learn More
Medchemexpress LLC Benzyloxycarbonyl-L-lysine N-hydroxysuccinimide ester | 21160-83-8 | MFCD00057964 | 95.0% | 511.52 g/mol | C26H29N3O8 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1295871G
Encompass_Preferred
Benzyloxycarbonyl-protected lysine N-hydroxysuccinimide ester is an activated amino acid derivative used as an NHS ester for formation of amide bonds with primary amines. It provides a Z (benzyloxycarbonyl) protecting group on the lysine side chain and is commonly used in peptide coupling and amine-derivatization workflows. Supplied as a powder for laboratory use.
  • Activated N-hydroxysuccinimide ester for efficient amide bond formation.
  • Provides benzyloxycarbonyl side-chain protection for lysine.
  • Suitable for peptide synthesis and amine derivatization.
  • Supplied as a powder with standard laboratory storage recommendations.
  • Compatible with common coupling methods for solid-phase and solution-phase synthesis.
Catalog No. 50-002-83432