Learn More
Medchemexpress LLC Benzyloxycarbonyl-LLY-fluoromethyl ketone | 133410-84-1 | 98.7% | C30H40FN3O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13675010MG
Encompass_Preferred
Z-LLY-FMK (benzyloxycarbonyl-LLY-fluoromethyl ketone) is a cell-permeant, irreversible inhibitor of calpain II used to investigate apoptosis and protease-mediated pathways in cell and animal models. It has demonstrated activity in cell-based assays (inhibiting Gal-9-induced apoptosis) and shown efficacy in animal studies by reducing hepatocyte apoptosis and parasite burden. Supplied as a high-purity solid for research use only.
  • Cell-permeant, irreversible calpain II inhibitor.
  • Validated in cell-based and in vivo apoptosis models.
  • High supplied purity (98.65%).
  • Supplied as a solid with defined storage recommendations.
  • Intended for research use only; not for human use.
Catalog No. 50-002-60994