Learn More
Medchemexpress LLC Benzyloxycarbonyl valacyclovir | 124832-31-1 | 98.09% | 458.47 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1263751G
Encompass_Preferred
Cbz-Valaciclovir (Benzyloxycarbonyl valacyclovir) is a chemical intermediate for antiviral research. It can be used to prepare Valaciclovir. This product is a white to off-white solid.
  • Can be used to prepare Valaciclovir
  • Useful for antiviral research
  • Solid appearance
  • White to off-white color
  • For research use only
Catalog No. 50-004-38042