Learn More
Medchemexpress LLC Benzyloxycarbonyl valacyclovir | 124832-31-1 | 98.1% | 458.47 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1263755G
Encompass_Preferred
Cbz-Valaciclovir (Benzyloxycarbonyl valacyclovir) is a compound that can be utilized in the preparation of Valaciclovir and for antiviral research. This product is intended for research use only and is not available for sale to patients.
  • Can be used to prepare Valaciclovir
  • Applicable for antiviral research
Catalog No. 50-004-35788