Learn More
Selleck Chemical LLC Bephenium Hydroxynaphthoate S4389-50mg
SDP
Supplier:  Selleck Chemical LLC S438950MG
Encompass_Preferred
Bephenium(NSC 759245) is an activator of B-type Acetylcholine receptor (AChR) Bephenium selectively activates the Hco-L-AChR1 subtype made of Hco-UNC-29 1 Hco-UNC-38 Hco-UNC-63 Hco-ACR-8 subunits Bephenium hydroxynaphthoate is an anthelmintic agent formerly used in the treatment of hookworm infections and ascariasis
Catalog No. 50-310-8193