Learn More
Apexbio Technology LLC Bephenium Hydroxynaphthoate 3818-50-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A83765.1
Encompass_Preferred
Bephenium Hydroxynaphthoate (CAS 3818-50-6) is an anthelmintic agonist targeting B-type acetylcholine receptors (AChR) It is designed to stimulate these receptors thereby disrupting neuromuscular transmission in gastrointestinal nematodes Bephenium Hydroxynaphthoate exerts its biological activity primarily through inhibition of parasite neuromuscular function causing paralysis and facilitating expulsion from the host intestine Experimental data on IC50 values are limited Based on these pharmacological properties Bephenium Hydroxynaphthoate holds research potential in studies of gastrointestinal nematode physiology parasite-host interactions and neuromuscular pharmacodynamics
Catalog No. 50-000-96070