Learn More
Medchemexpress LLC Bepotastine | 125602-71-3 | 98.6% | 388.89 | C21H25ClN2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYI002110MG
Encompass_Preferred
Bepotastine is a selective, orally active second-generation histamine H1 receptor antagonist supplied for laboratory research. It is provided in small milligram quantities and as solution formats for in vitro and preclinical studies. Chemical identifiers include CAS 125602-71-3, molecular formula C21H25ClN2O3, molecular weight 388.89 g/mol, and reported purity ~98.6%.
  • Selective H1 receptor antagonist for allergy-related research.
  • Suitable for studies of allergic rhinitis, conjunctivitis, and urticaria/pruritus.
  • Available as small milligram solid quantities and as DMSO solution formats.
  • High reported purity for reliable experimental results.
  • Includes standard chemical identifiers for traceability.
Catalog No. 50-002-97006