Learn More
Apexbio Technology LLC Bepotastine Besilate 190786-44-8 200mg
SDP
Supplier:  Apexbio Technology LLC B1569200
Encompass_Preferred
Bepotastine Besilate (CAS number 190786-44-8) is a small molecule antagonist targeting the histamine H1 receptor By binding selectively to this receptor subtype Bepotastine attenuates histamine-induced cellular responses decreasing symptoms associated with allergic reactions In experimental settings it serves as a pharmacological tool to investigate histamine-related inflammatory pathways and receptor signaling mechanisms Its utility extends to studies of allergic responses inflammation modulation and histamine receptor pharmacodynamics in biomedical research
Catalog No. 50-000-62004