Learn More
Apexbio Technology LLC Bepridil hydrochloride 68099-86-5 50mg
SDP
Supplier:  Apexbio Technology LLC B759150
Encompass_Preferred
Bepridil hydrochloride (CAS 68099-86-5) is a small molecule calcium channel blocker characterized by its ability to inhibit the influx of calcium ions into cardiac myocytes This modulation of calcium entry alters cardiac electrophysiology and contractility thereby supporting its use in angina pectoris research Bepridil hydrochloride has been investigated in both preclinical and clinical settings to explore its effects on cardiovascular pathophysiology mechanisms of arrhythmogenesis and potential as a therapeutic candidate It serves as a valuable tool in studies focused on cardiac ion channel regulation and the development of novel strategies for cardiovascular disease intervention
Catalog No. 50-000-88889