Learn More
Apexbio Technology LLC Berbamine hydrochloride 6078-17-7 1g
SDP
Supplier:  Apexbio Technology LLC N24711000
Encompass_Preferred
Berbamine hydrochloride (CAS 6078-17-7) is an isoquinoline alkaloid derivative isolated from plants of the Berberidaceae family It exerts its biological effects primarily through modulation of various signaling pathways including inhibition of STAT3 activation and disruption of calcium homeostasis in cells Berbamine hydrochloride has been investigated for its ability to suppress proliferation and induce apoptosis in multiple cancer cell types as well as for its immunomodulatory properties This compound serves as a valuable tool for studies exploring mechanisms of tumorigenesis and evaluating potential therapeutic strategies in cancer and immune-related research
Catalog No. 50-000-94451