Learn More
AdipoGen Berberine chloride
SDP
Supplier:  AdipoGen CDXB0212G050
Encompass_Preferred
Chemical. CAS 633-65-8. Formula C20H17NO4 . HCl. MW 335.3 . 36.5. Synthetic Natural fluorescent dye, used in histology for staining heparin in mast cells. An isoquinoline alkaloid, which shows antibacterial, antifungal, anti-inflammatory, anti-oxidant, antidiabetic, antiviral and anticancer activity. It inhibits cell growth and induces apoptosis in a variety of cancer cell lines. Broad spectrum enzyme inhibitor, affects N-acetyltransferase, cyclooxygenase-2, and topoisomerase activities and gene/protein expression. Increases adiponectin expression, upregulates LDLR and insulin receptor. Anti-hyperglycemic, by inhibition of aldose reductase, inducing glycolysis, preventing insulin resistance through increasing insulin receptor expression and acting like incretins. Activates AMPK and ERK. Potent, uncompetitive, reversible IDO 1 inhibitor with IC50 values of 7 µM cell-based assay and 9.3 µM enzymatic assay, and Ki value 8µM.
Catalog No. 50-256-1645