Learn More
Medchemexpress LLC Berberine chloride | 633-65-8 | 99.9% | 371.81 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1825810MM/1ML
Encompass_Preferred
Berberine chloride is an alkaloid derived from *Phellodendron amurense Rupr.*, classified as an isoquinoline alkaloid. It functions as an antibiotic by inducing reactive oxygen species (ROS) generation and inhibiting DNA topoisomerase. This compound also exhibits antineoplastic properties, making it a subject of interest in cancer research.
  • Acts as an antibiotic
  • Induces reactive oxygen species generation
  • Inhibits DNA topoisomerase
  • Possesses antineoplastic properties
  • Derived from *Phellodendron amurense Rupr.*
  • Classified as an isoquinoline alkaloid
  • Used in cancer research
Catalog No. 50-003-87626