Learn More
Medchemexpress LLC Berberine (chloride hydrate) | 68030-18-2 | 99.69% | 389.83 | 1 G
SDP
Supplier:  Medchemexpress LLC HY175771G
Encompass_Preferred
Berberine chloride hydrate is an alkaloid that acts as an antibiotic. It induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase. It also possesses antineoplastic properties.
  • Acts as an antibiotic.
  • Induces reactive oxygen species (ROS) generation.
  • Inhibits DNA topoisomerase.
  • Has antineoplastic properties.
  • Shows potential inhibitory effects on the proliferation of four colorectal carcinoma cell lines in vitro.
  • Induces a time- and dose-dependent inhibition of LoVo cell growth.
  • Exposure to 40 μM induces G2/M-phase cell cycle arrest in LoVo cells.
  • Suppresses cyclin B1, cdc2, and cdc25c protein expression in LoVo cells.
  • Inhibits the growth of human colorectal adenocarcinoma in vivo at doses of 30 and 50 mg/kg/day.
Catalog No. 50-003-87398