Learn More
Medchemexpress LLC Berberine (chloride hydrate) | 68030-18-2 | 99.82% | 389.83 | 5 G
SDP
Supplier:  Medchemexpress LLC HY175775G
Encompass_Preferred
Berberine chloride hydrate, also known as Natural Yellow 18 chloride hydrate, is an alkaloid with antibiotic properties. It induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase, demonstrating antineoplastic characteristics. This product is intended for research use only.
  • Acts as an antibiotic
  • Induces reactive oxygen species generation
  • Inhibits DNA topoisomerase
  • Exhibits antineoplastic properties
  • Shows potential inhibitory effects on proliferation of colorectal carcinoma cell lines
  • Induces time- and dose-dependent inhibition of cell growth
  • Leads to accumulation of cells in the G2/M phase
  • Suppresses protein expression
  • Inhibits human colorectal adenocarcinoma growth in vivo
Catalog No. 50-003-87142