Learn More
Medchemexpress LLC Berberine (chloride) (Standard) | 633-65-8 | 371.81 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1825810G
Encompass_Preferred
Berberine chloride is an alkaloid that acts as an antibiotic. It induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase, exhibiting antineoplastic properties. It is also known as Natural Yellow 18 chloride.
  • Alkaloid
  • Acts as an antibiotic
  • Induces reactive oxygen species (ROS) generation
  • Inhibits DNA topoisomerase
  • Has antineoplastic properties
  • Applicable in disease research fields, biological pesticide research, and agricultural research
Catalog No. 50-003-87712