Learn More
Medchemexpress LLC Berberine (hemisulfate) | 316-41-6 | 384.40 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0716A
Encompass_Preferred
Berberine hemisulfate, also known as Natural Yellow 18 hemisulfate, is an alkaloid derived from the traditional Chinese herbal medicine Huanglian. This compound is the hemisulfate form of Berberine.
  • Exhibits anti-inflammatory, antibiobic, antitumor, cardiovascular protective and neuroprotective activity.
  • Shows antimicrobial activity through inhibition of cell division protein FtsZ, or through DNA/RNA binding and subsequent DNA/RNA damage.
  • Exhibits anti-inflammatory activity by inhibiting TNF-α and the activation of its downstream pathway AP-1 and NF-κB.
  • Demonstrates neuroprotective efficacy by inhibiting reactive oxygen species (ROS) production and caspase activation, and activating the PI3K/Akt signaling pathway and heme oxygenase-1 (HO-1) expression.
  • Attenuates metabolic diseases through regulations of lipid composition and inhibition of insulin resistance.
Catalog No. 50-003-89174