Learn More
Apexbio Technology LLC Berberine hydrochloride 633-65-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N16995.1
Encompass_Preferred
Berberine hydrochloride (CAS 633-65-8) is a bioactive alkaloid designed to modulate glucose metabolism and related pathways Berberine hydrochloride exerts its biological activity primarily by inhibiting mitochondrial oxidative phosphorylation which promotes glycolysis and by functioning as an -glucosidase inhibitor thereby impeding disaccharidase activity and limiting intestinal glucose absorption In animal models with diet-induced obesity and clinical studies in type 2 diabetes patients berberine hydrochloride demonstrates enhanced glucose uptake increased glucose consumption under insulin-deficient conditions and improved insulin sensitivity No IC50 value has been reported Based on these pharmacological properties berberine hydrochloride holds research potential in diabetes and metabolic disorder applications
Catalog No. 50-000-90711