Learn More
Medchemexpress LLC Berberine sulfate | 633-66-9 | 433.43 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN0716B5G
Encompass_Preferred
Berberine sulfate, an alkaloid isolated from the Chinese herbal medicine Huanglian, functions as an antibiotic and exhibits antineoplastic properties. It induces reactive oxygen species (ROS) generation and inhibits DNA topoisomerase. The sulfate form is recognized for its improved bioavailability.
  • Isolated from Coptis chinensis Franch.
  • Acts as an antibiotic
  • Induces reactive oxygen species (ROS) generation
  • Inhibits DNA topoisomerase activity
  • Possesses antineoplastic properties
  • Improved bioavailability due to sulfate form
  • Purity of 99.23%
  • Inhibits proliferation of various colorectal carcinoma cell lines
  • Suppresses expression of key cell cycle proteins
  • Inhibits growth of human colorectal adenocarcinoma xenografts
Catalog No. 50-004-15470