Learn More
Medchemexpress LLC Berberine (sulfate) | 633-66-9 | 433.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0716B50MG
Encompass_Preferred
Berberine sulfate is an alkaloid isolated from the Chinese herbal medicine Huanglian, functioning as an antibiotic. It induces reactive oxygen species (ROS) generation, inhibits DNA topoisomerase, and possesses antineoplastic properties. The sulfate form of berberine is noted to improve its bioavailability.
  • An alkaloid isolated from the Chinese herbal medicine Huanglian.
  • Functions as an antibiotic.
  • Induces reactive oxygen species (ROS) generation.
  • Exhibits antineoplastic properties.
  • Inhibits DNA topoisomerase.
  • Sulfate form improves bioavailability.
Catalog No. 50-004-15891