Learn More
Medchemexpress LLC Berberine ursodeoxycholate | 1868138-66-2 | 99.3% | 727.93 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14724650MG
Encompass_Preferred
Berberine ursodeoxycholate (HTD1801) is an orally active and potent hypolipidemic agent, formed as an ionic salt of Berberine and Ursodeoxycholic acid. This compound has demonstrated a significant reduction in liver fat content and exhibits a broad spectrum of metabolic activity. It is suitable for research related to hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes.
  • Orally active and potent hypolipidemic agent.
  • Significantly reduces liver fat content.
  • Possesses a broad spectrum of metabolic activity.
  • Can be used in research for hyperlipidemia, non-alcoholic steatohepatitis (NASH), and diabetes.
  • Shows significant reductions in HbA1c (hemoglobin A1c) levels in vivo, with mean HbA1c levels decreasing by 0.6%.
  • Dissociates into Berberine and Ursodeoxycholic acid in the gastrointestinal tract, which are then absorbed differentially.
Catalog No. 50-003-62853