Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | 99.3% | 357.79 g/mol | C19H16ClNO4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12585010MG
Encompass_Preferred
Berberrubine chloride is the chloride salt of berberrubine, an orally active metabolite of berberine supplied for research use. It is provided as a solid reference standard for in vitro assays and preclinical studies, with reported anti-inflammatory and antitumor activities. Typical identifiers include CAS 15401-69-1, chemical formula C19H16ClNO4, molecular weight ~357.8 g/mol, and high analytical purity suitable for laboratory applications.
  • High purity, suitable for analytical and pharmacological studies.
  • Supplied as a stable solid reference standard for research use.
  • Documented chemical identity: CAS 15401-69-1 and formula C19H16ClNO4.
  • Reported biological activities include anti-inflammatory and antitumor effects.
  • Appropriate for in vitro assays and preclinical pharmacology models.
Catalog No. 50-002-39116