Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | 99.3% | 357.79 g·mol⁻¹ | C19H16ClNO4 | 5MG

Supplier: Medchemexpress LLC HY1258505MG
Berberrubine chloride is the chloride salt of berberrubine, an isoquinoline alkaloid and an orally active metabolite of berberine. It is provided as a research reagent for pharmacology and biological studies, including investigations of anti-inflammatory activity.
- CAS 15401-69-1.
- Chemical formula C19H16ClNO4; molecular weight 357.79 g·mol⁻¹.
- Purity 99.3% (reported by manufacturer).
- Available sizes include 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
- Storage sealed, away from moisture and light; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
- Intended for research use in in vitro and in vivo pharmacology assays, including anti-inflammatory models.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.