Learn More
Medchemexpress LLC Berberrubine chloride | 15401-69-1 | MFCD01658905 | 99.3% | 357.79 g·mol⁻¹ | C19H16ClNO4 | 100MG

Supplier: Medchemexpress LLC HY125850100MG
Berberrubine chloride is the chloride salt of berberrubine, a berberine metabolite with reported anti-inflammatory activity. This research-grade small molecule is supplied for laboratory use in milligram-scale quantities and is supported by quality documentation including a certificate of analysis and safety data sheet.
- High purity: 99.3%.
- Confirmed identity: CAS 15401-69-1.
- Chemical formula: C19H16ClNO4.
- Molecular weight: 357.79 g·mol⁻¹.
- Supplied in milligram-scale quantities, for example 100 MG vials.
- Intended for research use only; not for human or veterinary use.
- Documentation available: datasheet, COA, and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.