Learn More
Medchemexpress LLC HY-N2194 5mg Medchemexpress, Bergamottin CAS:7380-40-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21945MG
Encompass_Preferred
Medchemexpress, HY-N2194 5mg Bergamottin CAS:7380-40-7 Bergamottin is a potent and competitive CYP1A1 inhibitor with a K i of 10.703 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3894