Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bersiporocin | 2241808-52-4 | 99.0% | 328.24 g/mol | C15H19Cl2N3O | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1455555MG

Encompass_Preferred

Bersiporocin is a small-molecule prolyl-tRNA synthetase (PRS) inhibitor used in preclinical research to study antifibrotic mechanisms and collagen synthesis modulation. Supplied as a high-purity solid, it is suitable for in vitro and in vivo formulation with documented solubility in DMSO and recommended low-temperature storage for stability.

  • High purity (≈99%) suitable for research applications.
  • Potent PRS inhibition for antifibrotic research.
  • Soluble in DMSO at high concentration for in vitro studies.
  • Available in small mg quantities for early-stage experiments.
  • Stability data provided for low-temperature storage.

Catalog No. 50-002-21072


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.