Learn More
Medchemexpress LLC Beryllon II | 51550-25-5 | 98.0% | 738.52 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1122761G
Encompass_Preferred
Beryllon II is a widely used chromogenic reagent that is used to determine many elements, such as Mo, Mg, and Co, and is also used for the determination of proteins.
  • It has been used to determine many elements, such as Mo, Mg and Co.
  • The Beryllon II-Al3+ complex with the addition of protein can potently enhance Rayleigh light scattering, and is used to determine proteins.
Catalog No. 50-004-36617