Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BES sodium | 66992-27-6 | 97.0% | 235.23 | 500 G
SDP

Supplier:  Medchemexpress LLC 500GHYD0867A

Encompass_Preferred

BES sodium is a secondary standard biochemical buffer with an effective pH range of 6.4 to 7.8 (pKa: 7.15 at 20 °C). It is used in the diagnostic testing area and appears as a white to off-white solid. This product is for research use only.

  • Secondary standard biochemical buffer
  • Effective pH range of 6.4 to 7.8 (pKa: 7.15 at 20 °C)
  • Used in diagnostic testing
  • White to off-white solid
  • For research use only

Catalog No. 50-003-65844


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.