Learn More
Apexbio Technology LLC Besifloxacin HCl 405165-61-9 500mg
SDP
Supplier:  Apexbio Technology LLC B1895500
Encompass_Preferred
Besifloxacin HCl (CAS 405165-61-9) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to disrupt bacterial DNA replication and repair thereby inhibiting bacterial nucleic acid synthesis Besifloxacin HCl exerts its biological activity primarily through inhibition of DNA gyrase and topoisomerase IV In in vitro studies Besifloxacin HCl demonstrates inhibitory activity with IC50 values typically ranging from 0 1 to 0 8 g/mL depending on the specific bacterial strain tested Based on these pharmacological properties Besifloxacin HCl holds research potential in studies of antibacterial mechanisms resistance development and comparative antibiotic efficacy particularly in ocular infection models
Catalog No. 50-000-89858