Learn More
Medchemexpress LLC HY-B0134A 10mg Medchemexpress, Bestatin (hydrochloride) CAS:65391-42-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0134A10MG
Encompass_Preferred
Medchemexpress, HY-B0134A 10mg Bestatin (hydrochloride) CAS:65391-42-6 Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2375