Learn More
Expression Systems BESTBAC 1.0 LINEAR BACULOVIRUS
SDP
Supplier:  Expression Systems 91100
Encompass_Preferred
Expression Systems’ popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors. The resulting virus vector provides superior stability and purity compared to other systems.
• Unmatched productivity and stability for BEVS
• Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/ml of recombinant baculovirus in P1 stock
• Generates P1 stock as quickly as Bac-to-Bac® and other systems
• Each BestBac vial contains enough material for a minimum of five cotransfections
• Suitable for other expression systems including BacMam™
• BestBac 1.0 is wild type at the cathepsin and chitinase (v-cath/chiA) loci, preferred for some applications.
Included in the kit are:
• BestBac DNA
• pVL1393-lacZ control vector
• Transfection medium
• Expres2 TR transfection reagent
Catalog No. 50-165-6605