Learn More
Medchemexpress LLC Bet-in-19 | 1643947-30-1 | 99.7% | 333.39 | C19H19N5O | 5MG

Supplier: Medchemexpress LLC HY1252365MG
BET-IN-19 is a small-molecule BET bromodomain inhibitor used for preclinical research. It inhibits hIL-6 mRNA transcription and c-myc activity in human AML MV4-11 cells and disrupts tetra-acetylated histone H4 binding to BRD4 bromodomain 1. The compound is supplied as a solid and as DMSO solutions for biochemical and cellular studies.
- High purity (99.7%) suitable for research applications.
- Molecular formula C19H19N5O; molecular weight 333.39.
- CAS number 1643947-30-1 for unambiguous identification.
- Available as solid and concentrated DMSO solution formats.
- Recommended storage: powder -20°C; in solvent -80°C for long-term storage.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.