Learn More
Medchemexpress LLC Bet-in-19 | 1643947-30-1 | MFCD35200285 | 99.7% | 333.39 g/mol | C19H19N5O | 50MG
SDP
Supplier:  Medchemexpress LLC HY12523650MG
Encompass_Preferred
BET-IN-19 is a small-molecule BET bromodomain inhibitor (CAS 1643947-30-1) used in epigenetic and oncology research. It inhibits BRD4 bromodomain 1 binding to tetra-acetylated histone H4 and demonstrates cellular activity (reported IC50 ≤ 0.3 μM). Supplied as a powder with recommended storage at -20°C (powder) and in solvent at -80°C (6 months) or -20°C (1 month). Formula: C19H19N5O; molecular weight: 333.39 g/mol; reported purity ~99.7%.
  • Inhibits BRD4 bromodomain 1 with cellular activity (IC50 ≤ 0.3 μM).
  • High reported purity suitable for research applications.
  • Stable as a powder at -20°C for extended storage.
  • Solvent aliquots stable at -80°C for six months or -20°C for one month.
  • Offered in multiple pack sizes including 50 MG for convenience.
Catalog No. 50-002-37391